5-(Bromomethyl)-benzofurazan

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF516.2.5 CFF516.10 005888-2.5G
CHMPCFF516.2.5EA 0 GBP
CHMPCFF516.2.5 CHMPCFF516.10 FLUO005888-2.5G
5-(Bromomethyl)-benzofurazan
5-(Bromomethyl)-benzofurazan
Formula: C₇H₅BrN₂O
MDL Number: MFCD00174369
CAS Number: 32863-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR