Methyl-2-amino-4-(4-chlorophenyl)pyrimidine-5-carboxylate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2276-5G
HC-2276-1G
COBBHC-2276-5GEA
0
GBP
COBBHC-2276-5G
COBBHC-2276-1G
Methyl-2-amino-4-(4-chlorophenyl)pyrimidine-5-carboxylate
Methyl 2-amino-4-(4-chlorophenyl)pyrimidine-5-carboxylate
Formula:
C₁₂H₁₀ClN₃O₂ |
MDL Number:
MFCD11855874 CAS Number: 1133115-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...