1-Chloro-2,2-dimethylpropane ≥98%

Supplier: ABCR GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Chloro-1-diméthyl-2,2-propaneneo-Pentylchloride

ABCRAB119826.0100EA 0 GBP
ABCRAB119826.0100
1-Chloro-2,2-dimethylpropane ≥98%
1-Chloro-2,2-dimethylpropane
Formula: C₅H₁₁Cl
MW: 106.6 g/mol
Boiling Pt: 84…85 °C
Melting Pt: –20 °C
Density: 0.866
Flash Pt: 16 °F
MDL Number: MFCD00000941
CAS Number: 753-89-9
EINECS: 212-040-8
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR