Membrane filters, PORAFIL® CA
Supplier: Macherey-Nagel
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MANA68000080013EA 102 GBP
MANA68000080013 MANA68000080100 MANA68000080047 MANA68000080025 MANA68000045090 MANA68000020025 MANA68000020047 MANA68000045047 MANA68000045025 MANA68000045013 MANA68000020142 MANA68000020220 MANA68000020100 MANA68000020013 MANA68000080090 MANA68000020050 MANA68000045050 MANA68000080050 MANA68000020090 MANA68000045100 MANA68000045220 MANA68000045142 MANA68000080220 MANA68000080142
Membrane filters, PORAFIL® CA
Filters Membrane Filters
Cellulose acetate membranes have a low protein binding capacity and are suited for aqueous and alcoholic media.
  • Hydrophilic
  • Suitable for use with hot gases up to 180 °C
  • Sterilisable