(±)-α-Lipoic acid ≥99%, Calbiochem®, Sigma-Aldrich®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Lip(S2)6,8-Dithiooctanoic acid(±)​-​1,2-​Dithiolane-​3-pentanoic acid5-(1,2)Dithiolan-3-yl-pentanoic acidDL-Thioctic acid (oxidized)5-(1,2-Dithiolan-3-yl)pentanoic acidDL-α-Lipoic acidDL-6,8-Thioctic acid

M-Clarity™ quality level = MQ100

437692-5EA 77 GBP
437692-5
(±)-α-Lipoic acid ≥99%, Calbiochem®, Sigma-Aldrich®
(±)-α-Lipoic acid
Formula: C₈H₁₄O₂S₂
MW: 206.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005474
CAS Number: 1077-28-7
EINECS: 214-071-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR