(±)-α-Lipoic acid
Supplier: PanReac AppliChem
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Lip(S2)
, 6,8-Dithiooctanoic acid
, (±)-1,2-Dithiolane-3-pentanoic acid
, 5-(1,2)Dithiolan-3-yl-pentanoic acid
, DL-Thioctic acid (oxidized)
, 5-(1,2-Dithiolan-3-yl)pentanoic acid
, DL-α-Lipoic acid
, DL-6,8-Thioctic acid
A4065.0005EA
243
GBP
A4065.0005
(±)-α-Lipoic acid
(±)-α-Lipoic acid
Formula:
C₈H₁₄O₂S₂ MW: 206.33 g/mol |
MDL Number:
MFCD00005474 CAS Number: 1077-28-7 EINECS: 214-071-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...