2-(Methylamino)benzotrifluoride hydrochloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023289-25G 023289-5G
FLUO023289-25GEA 0 GBP
FLUO023289-25G FLUO023289-5G
2-(Methylamino)benzotrifluoride hydrochloride
2-(Methylamino)benzotrifluoride hydrochloride
Formula: C₈H₉ClF₃N
CAS Number: 845866-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR