2-Chloro-2'-nitroacetanilide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011117-25G 011117-100G 011117-5G
FLUO011117-25GEA 0 GBP
FLUO011117-25G FLUO011117-100G FLUO011117-5G
2-Chloro-2'-nitroacetanilide
2-Chloro-2'-nitroacetanilide
Formula: C₈H₇ClN₂O₃
MW: 214.61 g/mol
MDL Number: MFCD00024201
CAS Number: 10147-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR