7-Methoxybenzofuran-2-carboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018775-1G 018775-5G
FLUO018775-1GEA 0 GBP
FLUO018775-1G FLUO018775-5G
7-Methoxybenzofuran-2-carboxylic acid
7-Methoxybenzofuran-2-carboxylic acid
Formula: C₁₀H₈O₄
MW: 192.17 g/mol
MDL Number: MFCD00060516
CAS Number: 4790-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR