5-Methoxybenzofuran-2-carboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018774-1G 018774-5G
FLUO018774-1GEA 0 GBP
FLUO018774-1G FLUO018774-5G
5-Methoxybenzofuran-2-carboxylic acid
5-Methoxybenzofuran-2-carboxylic acid
Formula: C₁₀H₈O₄
MW: 192.1700 g/mol
MDL Number: MFCD00079771
CAS Number: 10242-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR