5-Chlorobenzofuran-2-carboxylic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Chlorobenzo[b]furan-2-carboxylic acid
018361-1G
018361-5G
FLUO018361-1GEA
0
GBP
FLUO018361-1G
FLUO018361-5G
5-Chlorobenzofuran-2-carboxylic acid
5-Chlorobenzofuran-2-carboxylic acid
Formula:
C₉H₅ClO₃ MW: 196.59 g/mol |
MDL Number:
MFCD00060512 CAS Number: 10242-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...