1-(2-Aminophenyl)pyrrole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(1H-Pyrrol-1-yl)aniline

013415-1G 013415-5G
FLUO013415-1GEA 0 GBP
FLUO013415-1G FLUO013415-5G
1-(2-Aminophenyl)pyrrole
1-(2-Aminophenyl)pyrrole
Formula: C₁₀H₁₀N₂
MW: 158.20 g/mol
MDL Number: MFCD00005344
CAS Number: 6025-60-1
EINECS: 227-884-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR