4-Iodo-D-phenylalanine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-D-Phe(4-I)-OH

M-Clarity™ quality level = MQ100

SIAL294462-500MGEA 303 GBP
SIAL294462-500MG
4-Iodo-D-phenylalanine, Sigma-Aldrich®
4-Iodo-D-phenylalanine
Formula: C₉H₁₀INO₂
MW: 291.1 g/mol
MDL Number: MFCD00063067
CAS Number: 62561-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR