4-Iodo-D-phenylalanine

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-D-Phe(4-I)-OH

BACE4010046.0250EA 0 GBP
BACE4010046.0250 BACE4010046.1000
4-Iodo-D-phenylalanine
4-Iodo-D-phenylalanine
Formula: C₉H₁₀INO₂
MW: 291.1 g/mol
MDL Number: MFCD00063067
CAS Number: 62561-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR