4-Chloro-L-phenylalanine

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: L-4-Chlorophenylalanine, H-Phe(4-Cl)-OH

4002990.0001 4002990.0005
BACE4002990.0001EA 0 GBP
BACE4002990.0001 BACE4002990.0005
4-Chloro-L-phenylalanine
4-Chloro-L-phenylalanine
Formula: C₉H₁₀ClNO₂
MW: 199.6 g/mol
MDL Number: MFCD00063065
CAS Number: 14173-39-8
EINECS: 238-023-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR