4-Bromo-L-phenylalanine

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-Phe(4-Br)-OH

BACE4007011.0001EA 80.99 GBP
BACE4007011.0001 BACE4007011.0005 BACE4007011.0025
4-Bromo-L-phenylalanine
4-Bromo-L-phenylalanine
Formula: C₉H₁₀BrNO₂
MW: 244.1 g/mol
MDL Number: MFCD00063062
CAS Number: 24250-84-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR