3-Fluoro-DL-phenylalanine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-3-(3-fluorophenyl)propionic acidH-DL-Phe(3-F)-OH

M-Clarity™ quality level = MQ200

SIALF5126-5GEA 409 GBP
SIALF5126-5G
3-Fluoro-DL-phenylalanine, Sigma-Aldrich®
3-Fluoro-DL-phenylalanine
Formula: C₉H₁₀FNO₂
MW: 183.2 g/mol
MDL Number: MFCD00004273
CAS Number: 2629-54-1
EINECS: 220-104-1
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR