Etidronic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Hydroxyethylidenebis(phosphonic acid)1-Hydroxyethylidenediphosphonic acid1-Hydroxyethylidene-1,1-diphosphonic acidHEDP1-Hydroxyethane-1,1-diphosphonic AcidOEDFKHDEPA

SIALH6773-100GEA 87.2 GBP
SIALH6773-100G SIALH6773-500G
Etidronic acid, Sigma-Aldrich®
Etidronic acid
Formula: C₂H₈O₇P₂
MW: 206.028 g/mol
MDL Number: MFCD00070585
CAS Number: 2809-21-4
EINECS: 220-552-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR