Etidronic acid ≥96%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1-Hydroxyethylidenebis(phosphonic acid)1-Hydroxyethylidenediphosphonic acid1-Hydroxyethylidene-1,1-diphosphonic acidHEDP1-Hydroxyethane-1,1-diphosphonic AcidOEDFKHDEPA

43502.10EA 0 GBP
43502.10 43502.09 43502.18 43502.25
Etidronic acid ≥96%
Etidronic acid
Formula: C₂H₈O₇P₂
MW: 206.028 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070585
CAS Number: 2809-21-4
EINECS: 220-552-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR