3',4'-(Methylenedioxy)acetophenone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

274801-25G 274801-5G
SIAL274801-25GEA 178 GBP
SIAL274801-25G SIAL274801-5G
3',4'-(Methylenedioxy)acetophenone, Sigma-Aldrich®
3',4'-(Methylenedioxy)acetophenone
Formula: C₉H₈O₃
MW: 164.16 g/mol
MDL Number: MFCD00005831
CAS Number: 3162-29-6
EINECS: 221-613-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR