Dimethyl allylphosphonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008035-1G 008035-5G
FLUO008035-1GEA 0 GBP
FLUO008035-1G FLUO008035-5G
Dimethyl allylphosphonate
Dimethyl allylphosphonate
Formula: C₅H₁₁O₃P
MDL Number: MFCD00014951
CAS Number: 757-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR