(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (R)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthalene, (R)-(+)-TolBINAP

M-Clarity™ quality level = MQ100

668966-1G 668966-250MG
SIAL668966-1GEA 123 GBP
SIAL668966-1G SIAL668966-250MG
(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, Sigma-Aldrich®
(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
Formula: C₄₈H₄₀P₂
MW: 678.78 g/mol
MDL Number: MFCD01311709
CAS Number: 99646-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR