Ethyl-2-naphthoate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E2280.10 E2280.50
XXXXE2280.10EA 0 GBP
XXXXE2280.10 XXXXE2280.50
Ethyl-2-naphthoate
Ethyl 2-naphthoate
Formula: C₁₃H₁₂O₂
MW: 200.23 g/mol
MDL Number: MFCD00016811
CAS Number: 3007-91-8
EINECS: 221-119-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR