Ethyl-1-naphthoate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E2270.10 E2270.50
XXXXE2270.10EA 0 GBP
XXXXE2270.10 XXXXE2270.50
Ethyl-1-naphthoate
Ethyl 1-naphthoate
Formula: C₁₃H₁₂O₂
MW: 200.24 g/mol
MDL Number: MFCD00016805
CAS Number: 3007-97-4
EINECS: 221-120-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR