4-Chloro-6-methyl-2-(trifluoromethyl)quinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2222-5G HC-2222-1G
COBBHC-2222-5GEA 0 GBP
COBBHC-2222-5G COBBHC-2222-1G
4-Chloro-6-methyl-2-(trifluoromethyl)quinoline
4-Chloro-6-methyl-2-(trifluoromethyl)quinoline
Formula: C₁₁H₇ClF₃N
MW: 245.630 g/mol
MDL Number: MFCD00153097
CAS Number: 1701-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR