8-Bromo-5-(trifluoromethyl)quinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6997-1G HC-6997-5G
COBBHC-6997-1GEA 0 GBP
COBBHC-6997-1G COBBHC-6997-5G
8-Bromo-5-(trifluoromethyl)quinoline
8-Bromo-5-(trifluoromethyl)quinoline
Formula: C₁₀H₅BrF₃N
MDL Number: MFCD11504935
CAS Number: 917251-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR