trans-4-Hydroxy-β-nitrostyrene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018725-2G 018725-5G
FLUO018725-2GEA 0 GBP
FLUO018725-2G FLUO018725-5G
trans-4-Hydroxy-β-nitrostyrene
trans-4-Hydroxy-β-nitrostyrene
Formula: C₈H₇NO₃
MW: 165.1461 g/mol
MDL Number: MFCD00060551
CAS Number: 22568-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR