1,2-Dibromopentafluoropropyl-2,2,2-trifluoroethyl ether

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010288-1G 010288-5G
FLUO010288-1GEA 0 GBP
FLUO010288-1G FLUO010288-5G
1,2-Dibromopentafluoropropyl-2,2,2-trifluoroethyl ether
1,2-Dibromopentafluoropropyl-2,2,2-trifluoroethyl ether
Formula: C₅H₂Br₂F₈O
MDL Number: MFCD03094229
CAS Number: 396716-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR