5-Formyl-2-furanylboronic acid ≥95%, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Formylfuran-5-boronic acid, (5-formylfuran-2-yl)boronic acid, 5-formylfuran-2-boronic acid, (5-Formylfuran-2-yl)boronic acid, 5-Formyl-2-furanboronic acid

M-Clarity™ quality level = MQ100

512346-1G 512346-5G
SIAL512346-1GEA 37.2 GBP
SIAL512346-1G SIAL512346-5G
5-Formyl-2-furanylboronic acid ≥95%, Sigma-Aldrich®
5-Formyl-2-furanylboronic acid
Formula: C₅H₅BO₄
MW: 139.90 g/mol
MDL Number: MFCD01114696
CAS Number: 27329-70-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR