5-Formyl-2-furanylboronic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Formylfuran-5-boronic acid, (5-formylfuran-2-yl)boronic acid, 5-formylfuran-2-boronic acid, (5-Formylfuran-2-yl)boronic acid, 5-Formyl-2-furanboronic acid
011036-5G
011036-1G
FLUO011036-5GEA
0
GBP
FLUO011036-5G
FLUO011036-1G
5-Formyl-2-furanylboronic acid
5-Formyl-2-furanylboronic acid
Formula:
C₅H₅BO₄ MW: 139.90 g/mol |
MDL Number:
MFCD01114696 CAS Number: 27329-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...