5-Formyl-2-furanylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Formylfuran-5-boronic acid, (5-formylfuran-2-yl)boronic acid, 5-formylfuran-2-boronic acid, (5-Formylfuran-2-yl)boronic acid, 5-Formyl-2-furanboronic acid

BB-2298-5G BB-2298-25G BB-2298-1G BB-2298-100G
COBBBB-2298-5GEA 0 GBP
COBBBB-2298-5G COBBBB-2298-25G COBBBB-2298-1G COBBBB-2298-100G
5-Formyl-2-furanylboronic acid
5-Formyl-2-furanylboronic acid
Formula: C₅H₅BO₄
MW: 139.90 g/mol
MDL Number: MFCD01114696
CAS Number: 27329-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR