4-Formylfuran-2-boronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2296-1G BB-2296-250M
COBBBB-2296-1GEA 0 GBP
COBBBB-2296-1G COBBBB-2296-250M
4-Formylfuran-2-boronic acid
4-Formylfuran-2-boronic acid
Formula: C₅H₅BO₄
MDL Number: MFCD02179485
CAS Number: 62306-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR