3,4-(Methylenedioxy)phenylacetic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(1,3-Benzodioxol-5-yl)acetic acid
018839-5G
018839-25G
FLUO018839-5GEA
0
GBP
FLUO018839-5G
FLUO018839-25G
3,4-(Methylenedioxy)phenylacetic acid
3,4-(Methylenedioxy)phenylacetic acid
Formula:
C₉H₈O₄ MW: 180.16 g/mol |
MDL Number:
MFCD00014576 CAS Number: 2861-28-1 EINECS: 220-679-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...