2,2-Difluoro-1,3-benzodioxole-4-carbonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005736-1G 005736-5G
FLUO005736-1GEA 0 GBP
FLUO005736-1G FLUO005736-5G
2,2-Difluoro-1,3-benzodioxole-4-carbonyl chloride
2,2-Difluoro-1,3-benzodioxole-4-carbonyl chloride
Formula: C₈H₃ClF₂O₃
MDL Number: MFCD01631474
CAS Number: 143096-86-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR