2,5-Bis(trifluoromethyl)phenylacetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013767-1G 013767-5G
FLUO013767-1GEA 0 GBP
FLUO013767-1G FLUO013767-5G
2,5-Bis(trifluoromethyl)phenylacetonitrile
2,5-Bis(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₅F₆N
CAS Number: 302911-99-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR