4-Chloro-8-(trifluoromethyl)quinoline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF993.1 004562-1G 004562-5G
CHMPCFF993.1EA 0 GBP
CHMPCFF993.1 FLUO004562-1G FLUO004562-5G
4-Chloro-8-(trifluoromethyl)quinoline
4-Chloro-8-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MW: 231.61 g/mol
MDL Number: MFCD00134578
CAS Number: 23779-97-7
EINECS: 245-880-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR