4-Chloro-8-(trifluoromethyl)quinoline ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
A17301.03
A17301.06
A17301.03EA
37.1
GBP
A17301.03
A17301.06
4-Chloro-8-(trifluoromethyl)quinoline ≥98%
4-Chloro-8-(trifluoromethyl)quinoline
Purity: 98%
CAS No.: 23779-97-7
Molecular Formula: C10H5ClF3N
Formula Weight: 231.61
Melting Point: 80-82°
MDL No.: MFCD00134578
Formula:
C₁₀H₅ClF₃N MW: 231.61 g/mol Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134578 CAS Number: 23779-97-7 EINECS: 245-880-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...