4-Chloro-6-fluoro-2-(trifluoromethyl)quinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2170-5G HC-2170-1G
COBBHC-2170-5GEA 0 GBP
COBBHC-2170-5G COBBHC-2170-1G
4-Chloro-6-fluoro-2-(trifluoromethyl)quinoline
4-Chloro-6-fluoro-2-(trifluoromethyl)quinoline
Formula: C₁₀H₄ClF₄N
MDL Number: MFCD00276579
CAS Number: 59611-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR