1-(2-Thienyl)-1-propanone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Propionylthiophene

001219-100G 001219-25G
FLUO001219-100GEA 0 GBP
FLUO001219-100G FLUO001219-25G
1-(2-Thienyl)-1-propanone
1-(2-Thienyl)-1-propanone
Formula: C₇H₈OS
MW: 140.20 g/mol
MDL Number: MFCD00005446
CAS Number: 13679-75-9
EINECS: 237-182-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR