1-(Methoxymethyl)-3-(trifluoromethyl)benzene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0769-5G
OT-0769-1G
COBBOT-0769-5GEA
0
GBP
COBBOT-0769-5G
COBBOT-0769-1G
1-(Methoxymethyl)-3-(trifluoromethyl)benzene
1-(Methoxymethyl)-3-(trifluoromethyl)benzene
Formula:
C₉H₉F₃O |
MDL Number:
MFCD11504958 CAS Number: 380633-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...