(2,6-Difluor-3-methylphenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005763-1G 005763-2.5G
FLUO005763-1GEA 0 GBP
FLUO005763-1G FLUO005763-2.5G
(2,6-Difluor-3-methylphenyl)acetic acid
(2,6-Difluor-3-methylphenyl)acetic acid
Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
MDL Number: MFCD01631338
CAS Number: 261943-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR