3-(Trifluoromethyl)phenylacetone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-[3-(Trifluoromethyl)phenyl]-2-propanone

009454-5G 009454-25G
FLUO009454-5GEA 0 GBP
FLUO009454-5G FLUO009454-25G
3-(Trifluoromethyl)phenylacetone
3-(Trifluoromethyl)phenylacetone
Formula: C₁₀H₉F₃O
MW: 202.18 g/mol
MDL Number: MFCD00000397
CAS Number: 21906-39-8
EINECS: 244-652-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR