2-Chloro-3-nitropyridine ≥99.0% (by GC)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
C1008-25G
C1008-5G
TCIAC1008-25GEA
0
GBP
TCIAC1008-25G
TCIAC1008-5G
2-Chloro-3-nitropyridine ≥99.0% (by GC)
2-Chloro-3-nitropyridine
Formula:
C₅H₃ClN₂O₂ MW: 158.54 g/mol Boiling Pt: 260 °C (1013 hPa) Melting Pt: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006232 CAS Number: 5470-18-8 EINECS: 226-799-8 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 101.0 to 104.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...