Bromocyclobutane ≥96.0% (by GC)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Cyclobutyl bromide
C0771-1G
C0771-5G
TCIAC0771-1GEA
0
GBP
TCIAC0771-1G
TCIAC0771-5G
Bromocyclobutane ≥96.0% (by GC)
Bromocyclobutane
Formula:
C₄H₇Br MW: 135 g/mol Boiling Pt: 108°C Density: 1,4208 g/cm³ (20 °C) Flash Pt: 22°C |
MDL Number:
MFCD00001317 CAS Number: 4399-47-7 EINECS: 224-530-9 UN: 1993 ADR: 3,II |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...