2,6-Xylidine ≥99.0% (by GC)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Amino-m-xylene, 2,6-Dimethylaniline
D0669-25ML
D0669-500ML
TCIAD0669-25MLEA
0
GBP
TCIAD0669-25ML
TCIAD0669-500ML
2,6-Xylidine ≥99.0% (by GC)
2,6-Xylidine
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 216°C Melting Pt: 10…12 °C Density: 0,98 g/cm³ (20 °C) Flash Pt: 91°C Storage Temperature: Ambient |
MDL Number:
MFCD00007747 CAS Number: 87-62-7 EINECS: 201-758-7 UN: 1711 ADR: 6.1,II Merck Index: 12,10217 |
Specification Test Results
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...