2-Bromo-6-methoxynaphthalene ≥98.0% (by GC)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
B1547-25G
B1547-250G
TCIAB1547-25GEA
0
GBP
TCIAB1547-25G
TCIAB1547-250G
2-Bromo-6-methoxynaphthalene ≥98.0% (by GC)
2-Bromo-6-methoxynaphthalene
Formula:
C₁₁H₉BrO MW: 237.1 g/mol Boiling Pt: 164°C |
MDL Number:
MFCD00004062 CAS Number: 5111-65-9 EINECS: 225-837-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 107.0 to 111.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...