2,6-Dimethoxyphenol ≥98.0% (by GC)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
D0639-25G
D0639-500G
TCIAD0639-25GEA
0
GBP
TCIAD0639-25G
TCIAD0639-500G
2,6-Dimethoxyphenol ≥98.0% (by GC)
2,6-Dimethoxyphenol
Formula:
C₈H₁₀O₃ MW: 154.17 g/mol Boiling Pt: 263°C Melting Pt: 52…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064434 CAS Number: 91-10-1 EINECS: 202-041-1 |
Specification Test Results
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 54.0 to 57.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...