3-Iodobenzonitrile

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBOR-0932-25GEA 0 GBP
COBBOR-0932-25G
3-Iodobenzonitrile
3-Iodobenzonitrile
Formula: C₇H₄IN
MW: 229.02 g/mol
MDL Number: MFCD00079762
CAS Number: 69113-59-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR