2,6-Dichlorophenylhydrazinium chloride

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,6-Dichlorophenylhydrazine hydrochloride1-(2,6-Dichlorophenyl)hydrazine hydrochloride

COBBOS-7730-5GEA 0 GBP
COBBOS-7730-5G
2,6-Dichlorophenylhydrazinium chloride
2,6-Dichlorophenylhydrazinium chloride
Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
MDL Number: MFCD00012930
CAS Number: 50709-36-9
EINECS: 256-730-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR