2'-Hydroxy-4',6'-dimethoxyacetophenone ≥98.0% (by GC)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-(2-Hydroxy-4,6-dimethoxyphenyl)ethan-1-one, Xanthoxylin
D2683-1G
D2683-5G
TCIAD2683-1GEA
0
GBP
TCIAD2683-1G
TCIAD2683-5G
2'-Hydroxy-4',6'-dimethoxyacetophenone ≥98.0% (by GC)
2'-Hydroxy-4',6'-dimethoxyacetophenone
Formula:
C₁₀H₁₂O₄ MW: 196.2 g/mol Boiling Pt: 185°C |
MDL Number:
MFCD00017243 CAS Number: 90-24-4 EINECS: 201-978-3 Merck Index: 14,10067 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 79.0 to 82.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...